missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ S-CLASSIC-90 2-Door Light Gray Safety Storage Cabinets with Door Open Arrest System
For the safe and approved storage of flammable hazardous substances in working areas, with an optional door open arrest system.
Brand: Asecos™ 3020500133705
Code : NEW
Additional Details : Weight : 60.00000kg
Description
- Aseco’s first EN-compliant safety storage cabinet
- Decades of developing and optimizing ensure that the S-CLASSIC-90 is technically sophisticated
- Available in a range of widths and heights
- Versatile fields of application due to various interior equipment
- Convenient handling – door operation with minimal effort
- Optional door open arrest system allows convenient and safe removal or storage of containers with both hands
- Door lockable with cylinder lock, the cabinets can be integrated by the customer in an existing locking system
- Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- Fire resistance of 90 minutes
- Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- In accordance with EN 14470-1 and TRGS 510
Specifications
Door open arrest system | |
371 kg | |
Safety storage cabinet | |
3x 75 kg sheet steel shelves; 1x 75 kg sheet steel insert; 1x 22 L sheet steel sump | |
Powder-coated | |
2 |
616 x 896 x 1968 mm | |
Light Gray | |
Storage of flammable and hazardous substances | |
Triple-hinged | |
Steel | |
EN 14470-1 |