Learn More
Thermo Scientific Alfa Aesar Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated, 99.8% (metals basis)
Brand: Thermo Scientific Alfa Aesar 046365.RF
Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated, 99.8% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-50-8 Weight : 0.01000kg