missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Calcium fluoride crystal optic disc, 32mm x 3mm (drilled), polished both sides

Brand:  Thermo Scientific Alfa Aesar 038432.KT

Calcium fluoride crystal optic disc, 32mm x 3mm (drilled), polished both sides

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15406235

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.