Learn More
SI Analytics TITRONIC 300 Module 1 Basic Unit
Robust, reliable, ready-to-use, TITRONIC 300, Module 1 basic unit that is ideal for use in many laboratory and scientific and research environments.
Brand:  SI Analytics 285225810
Code : NEW
Additional Details : Weight : 5.00000kg
Product Code. 15354408
  • £1695.00 / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
  • TITRONIC 300 Module 1 TM 300/50 100-240V
  • Motor-driven piston burette with 20 or 50 ml dosing unit
  • For manual titrations and highly precise dosing tasks
  • The TITRONIC ™ 300 can completely controlled from a PC via the USB- or RS-232-interface
  • Up to 16 burettes can be connected to one another via the RS232 interface according to the daisy chain principle
Specifications
Specifications
TITRONIC 300 Module 1 Basic Unit Titrator
100-240V