missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Attune™ NxT Dichroic Filter, 495DLP
Click to view available options
Quantity:
1 filter
Unit Size:
Each
Description
- Replacement dichroic filter
- Used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer
Specifications
Specifications
| Product Line | Attune |
| Content And Storage | Store at room temperature. |
| Quantity | 1 filter |
| For Use With (Equipment) | Attune Acoustic Focusing Cytometer |
| Product Type | Dichroic Filter |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction