missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ 15-PGDH/HPGD Antibody Pair [HRP]

Product Code. p-7183237 Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Quantity:
15 Plates
5 Plates
missing translation for 'unitSize'
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18257476

missing translation for 'mfr': Novus Biologicals™ NBP27961515PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human 15-PGDH/HPGD.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 31.25-2000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human 15-PGDH/HPGD.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias 15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1
Gene ID (Entrez) 3248
Gene Symbol HPGD
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human 15-PGDH/HPGD monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of rabbit anti-Human 15-PGDH/HPGD monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 67 ng of recombinant Human 15-PGDH/HPGD.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Immunology, Innate Immunity
    Storage Requirements Storage is content dependent.
    Target 15-PGDH/HPGD
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.