missing translation for 'onlineSavingsMsg'
Learn More
Learn More
15-PGDH/HPGD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-87062-25ul
This item is not returnable.
View return policy
Description
15-PGDH/HPGD Polyclonal specifically detects 15-PGDH/HPGD in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| 15-PGDH/HPGD | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| 15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human 15-PGDH/HPGD antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| HPGD | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VDWNLEAGVQCKAALDEQFEPQKTLFIQCDVADQQQLRDTFRKVVDHFGRLDILVNNAGVNNEKNWEKTLQINLVSVISGTYLGLDYMSKQNGGEGGIIINMSSLAGLMPVAQQPV | |
| 25 μL | |
| Cancer, Immunology, Innate Immunity | |
| 3248 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction