missing translation for 'onlineSavingsMsg'
Learn More
Learn More
15-PGDH/HPGD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98291-50ul
This item is not returnable.
View return policy
Description
15-PGDH/HPGD Polyclonal antibody specifically detects 15-PGDH/HPGD in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| 15-PGDH/HPGD | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 μL/mg of lysate | |
| 15-hydroxyprostaglandin dehydrogenase [NAD+], 15-PGDH, EC 1.1.1, EC 1.1.1.141, hydroxyprostaglandin dehydrogenase 15-(NAD), NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, PGDH1PGDH, Prostaglandin dehydrogenase 1, SDR36C1, short chain dehydrogenase/reductase family 36C, member 1 | |
| Produced in rabbits immunized with purified, recombinant Mouse 15-PGDH/HPGD (Uniprot#: Q8VCC1; Met 1-Ser 269). | |
| 50 μg | |
| Cancer, Immunology, Innate Immunity | |
| 3248 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction