missing translation for 'onlineSavingsMsg'
Learn More

15-Lipoxygenase 2 Antibody, Novus Biologicals™

Product Code. 18382011 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382011 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382011 Supplier Novus Biologicals Supplier No. NBP286924

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

15-Lipoxygenase 2 Polyclonal specifically detects 15-Lipoxygenase 2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 15-Lipoxygenase 2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias 15-lipoxygenase 2, 15-LOX-B, arachidonate 15-lipoxygenase 2, arachidonate 15-lipoxygenase B, Arachidonate 15-lipoxygenase type II, arachidonate 15-lipoxygenase, second type, arachidonate 15-lipoxygenase, type B, arachidonate omega(6) lipoxygenase, EC 1.13.11, EC 1.13.11.33,15-LOX-215S-lipoxygenase
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of human 15-Lipoxygenase 2
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 247
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.