missing translation for 'onlineSavingsMsg'
Learn More
Learn More
15-Lipoxygenase 1 Antibody (OTI3G8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70169CL1
This item is not returnable.
View return policy
Description
15-Lipoxygenase 1 Monoclonal antibody specifically detects 15-Lipoxygenase 1 in Canine,Human,Monkey,Primate,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| 15-Lipoxygenase 1 | |
| Monoclonal | |
| CoraFluor 1 | |
| 15-lipooxygenase-1, 15-LOX-1, 15LOX-1, arachidonate 15-lipoxygenase, Arachidonate omega-6 lipoxygenase, EC 1.13.11, EC 1.13.11.33, LOG15,15-LOX | |
| Full length human recombinant protein of human ALOX15(NP_001131) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 246 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI3G8 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Primate, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction