missing translation for 'onlineSavingsMsg'
Learn More

15-Keto Travoprost, TRC

Product Code. 30133556
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133556 25 mg 25mg
30009623 10 mg 10mg
30011141 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133556 Supplier Toronto Research Chemicals Supplier No. TRCK31520025MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 404830-45-1
Molecular Formula C26 H33 F3 O6
Molecular Weight (g/mol) 498.53
Synonym (5Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(1E)-3-oxo-4-[3-(trifluoromethyl)phenoxy]-1-buten-1-yl]cyclopentyl]-5-heptenoic Acid 1-Methylethyl Ester, 15-Keto Travoprost
IUPAC Name propan-2-yl (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(E)-3-oxo-4-[3-(trifluoromethyl)phenoxy]but-1-enyl]cyclopentyl]hept-5-enoate
SMILES CC(C)OC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\C(=O)COc2cccc(c2)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C26H33F3O6/c1-17(2)35-25(33)11-6-4-3-5-10-21-22(24(32)15-23(21)31)13-12-19(30)16-34-20-9-7-8-18(14-20)26(27,28)29/h3,5,7-9,12-14,17,21-24,31-32H,4,6,10-11,15-16H2,1-2H3/b5-3-,13-12+/t21-,22-,23+,24-/m1/s1
Formula Weight 498.2229
Chemical Name or Material 15-Keto Travoprost
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.