missing translation for 'onlineSavingsMsg'
Learn More

15-Keto Latanoprost, TRC

Product Code. 30004878
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004878 10 mg 10mg
30004293 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004878 Supplier Toronto Research Chemicals Supplier No. TRCK19535010MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135646-98-9
Molecular Formula C23 H30 O5
Molecular Weight (g/mol) 386.48
Synonym 5-Heptenoic acid, 7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-(3-oxo-5-phenylpentyl)cyclopentyl]-, 1-methylethyl ester, (5Z)-, 5-Heptenoic acid, 7-[3,5-dihydroxy-2-(3-oxo-5-phenylpentyl)cyclopentyl]-, 1-methylethyl ester, [1R-[1α(Z),2β,3α,5α]]-, 15-Ketolatanoprost, 15-Keto Latanoprost
IUPAC Name (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(E)-3-oxo-5-phenylpent-1-enyl]cyclopentyl]hept-5-enoic acid
SMILES O[C@@H]1C[C@H](O)[C@H](C\C=C/CCCC(=O)O)[C@H]1\C=C\C(=O)CCc2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H30O5/c24-18(13-12-17-8-4-3-5-9-17)14-15-20-19(21(25)16-22(20)26)10-6-1-2-7-11-23(27)28/h1,3-6,8-9,14-15,19-22,25-26H,2,7,10-13,16H2,(H,27,28)/b6-1-,15-14+/t19-,20-,21+,22-/m1/s1
Formula Weight 386.2093
Chemical Name or Material 15-Keto Latanoprost
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.