missing translation for 'onlineSavingsMsg'
Learn More

15-Keto Bimatoprost, TRC

Product Code. 30010008
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010008 1 mg 1mg
30007949 25 mg 25mg
30003509 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010008 Supplier Toronto Research Chemicals Supplier No. TRCK1751001MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1163135-96-3
Molecular Formula C25 H35 N O4
Molecular Weight (g/mol) 413.55
Synonym 5-Heptenamide, 7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E)-3-oxo-5-phenyl-1-penten-1-yl]cyclopentyl]-N-ethyl-, (5Z)-, (5Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(1E)-3-oxo-5-phenyl-1-penten-1-yl]cyclopentyl]-N-ethyl-5-heptenamide, 15-Ketobimatoprost
IUPAC Name (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(E)-3-oxo-5-phenylpent-1-enyl]cyclopentyl]-N-ethylhept-5-enamide
SMILES CCNC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\C(=O)CCc2ccccc2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C25H35NO4/c1-2-26-25(30)13-9-4-3-8-12-21-22(24(29)18-23(21)28)17-16-20(27)15-14-19-10-6-5-7-11-19/h3,5-8,10-11,16-17,21-24,28-29H,2,4,9,12-15,18H2,1H3,(H,26,30)/b8-3-,17-16+/t21-,22-,23+,24-/m1/s1
Formula Weight 413.2566
Percent Purity >95%
Chemical Name or Material 15-Ketobimatoprost
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.