missing translation for 'onlineSavingsMsg'
Learn More

15-Hydroxypentadecanoic Acid, TRC

Product Code. 30400121
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400121 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400121 Supplier Toronto Research Chemicals Supplier No. TRCH296755100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4617-33-8
Molecular Formula C15H30O3
Quantity 100 mg
Synonym 14-Hydroxy-1-tetradecanecarboxylic Acid, ω-Hydroxypentadecanoic Acid
InChI Formula InChI=1S/C15H30O3/c16-14-12-10-8-6-4-2-1-3-5-7-9-11-13-15(17)18/h16H,1-14H2,(H,17,18)
SMILES OCCCCCCCCCCCCCCC(=O)O
IUPAC Name 15-hydroxypentadecanoic acid
Molecular Weight (g/mol) 258.4
Formula Weight 258.2195
Chemical Name or Material 15-Hydroxypentadecanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.