missing translation for 'onlineSavingsMsg'
Learn More

15-Hydroxypentadecanoic acid, 99+%

Product Code. 11470247
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470247 1 g 1g
11480247 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470247 Supplier Thermo Scientific Alfa Aesar Supplier No. B21670.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4617-33-8 | C15H30O3 | 258.402 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4617-33-8
Molecular Formula C15H30O3
Molecular Weight (g/mol) 258.402
MDL Number MFCD00039535
InChI Key BZUNJUAMQZRJIP-UHFFFAOYSA-N
PubChem CID 78360
ChEBI CHEBI:79169
IUPAC Name 15-hydroxypentadecanoic acid
SMILES C(CCCCCCCO)CCCCCCC(=O)O

Specifications

Melting Point 84°C to 88°C
Linear Formula HO(CH2)14CO2H
Quantity 1 g
Beilstein 1240100
Synonym 15-hydroxy-pentadecanoic acid, pentadecanoic acid, 15-hydroxy, acmc-1anlj, pentadecanoic acid,15-hydroxy, 15-hydroxypentadecanoic acid, 15-hydroxypentadecanoic acid capillary gc
Formula Weight 258.4
Percent Purity ≥99%
Chemical Name or Material 15-Hydroxypentadecanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.