missing translation for 'onlineSavingsMsg'
Learn More

15-Hydroxy Lubiprostone Dicyclohexylammonium Salt-d7, TRC

Product Code. 30391548
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391548 1 mg 1mg
30402792 0.25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391548 Supplier Toronto Research Chemicals Supplier No. TRCH9438921MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C32H50D7F2NO5
Molecular Weight (g/mol) 580.84
Synonym (11α)-16,16-Difluoro-11,15-dihydroxy-9-oxoprostan-1-oic Acid-d7, Dicyclohexylammonium Salt, 13,14-Dihydro-16,16-difluoroprostaglandin E1-d7 Dicyclohexylammonium Salt
IUPAC Name dicyclohexylamine 7-((1R,2R,3R)-2-(4,4-difluoro-3-hydroxyoctyl-6,6,7,7,8,8,8-d7)-3-hydroxy-5-oxocyclopentyl)heptanoate
SMILES FC(CC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(F)C(O)CC[C@@H]1[C@@H](CCCCCCC(O)=O)C(C[C@H]1O)=O.C2(NC3CCCCC3)CCCCC2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H34F2O5.C12H23N/c1-2-3-12-20(21,22)18(25)11-10-15-14(16(23)13-17(15)24)8-6-4-5-7-9-19(26)27;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h14-15,17-18,24-25H,2-13H2,1H3,(H,26,27);11-13H,1-10H2/t14-,15-,17-,18?;/m1./s1/i1D3,2D2,3D2;
Formula Weight 580.46
Chemical Name or Material 15-Hydroxy Lubiprostone Dicyclohexylammonium Salt-d7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.