missing translation for 'onlineSavingsMsg'
Learn More
Learn More
15% Biodiesel/85% High Cetane Diesel Fuel, v/v, VHG™
Description
15% Biodiesel/85% High Cetane Diesel Fuel, v/v
Specifications
Specifications
| Chemical Name or Material | Bio Diesel Fuel |
| CAS | 870530-78-2 |
| Analyte or Component Names | Bio Diesel Fuel |
| Shipping Condition | Room Temperature |
| Concentration | 15% |
| Synonym | Diesel fuel substitutes, biodiesel, B 10, B 10 biofuel, B 100, B 100 biofuel, B 20, B 20 biofuel, B 5, B 5 biofuel, B 50, B 50 biofuel, Biodiesel, Biofuels, biodiesel, Biodiesel fuel |
| IUPAC Name | Biodiesel Fuel |
| Solvent or Matrix | High Cetane Diesel Fuel/Biodiesel |
| Quantity | 20 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?