missing translation for 'onlineSavingsMsg'
Learn More

15-Azido-4,7,10,13-tetraoxapentadecanoic acid

Product Code. 15486159
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15486159

Brand: Thermo Scientific Alfa Aesar J64030.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1257063-35-6 | C11H21N3O6 | 291.30 g/mol

Azido-PEG4-Acid is amine- or phosphine-reactive building block used to derivative primary amines for ligation to alkyne-tagged molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azido-PEG4-Acid is amine- or phosphine-reactive building block used to derivative primary amines for ligation to alkyne-tagged molecules.

Solubility
Soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257063-35-6
Molecular Formula C11H21N3O6
Molecular Weight (g/mol) 291.30
MDL Number MFCD12406155
InChI Key BODPHGOBXPGJKO-UHFFFAOYSA-N
Synonym azido-peg4-acid, 15-azido-4,7,10,13-tetraoxapentadecanoic acid, n3-peg 4-cooh, 1-azido-3,6,9,12-tetraoxapentadecan-15-oic acid, azido-dpeg r 4-acid, 3-2-2-2-2-azidoethoxy ethoxy ethoxy ethoxy propanoic acid
PubChem CID 22731907

Specifications

Color Colorless to Yellow
Linear Formula N3CH2CH2(OCH2CH2)4CO2H
Quantity 100 mg
Solubility Information Soluble in water.
Formula Weight 291.3
Physical Form Liquid
Chemical Name or Material 15-Azido-4,7,10,13-tetraoxapentadecanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.