missing translation for 'onlineSavingsMsg'
Learn More

(14bRS)-2-Methyl-1,3,4,14b-tetrahydropyrazino[2,1-a]pyrido-[2,3-c][2]benzazepin-10(2H)-one (10-Oxomirtazapine), Mikromol™

Product Code. 30051609
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051609 100 mg 100mg
30050913 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051609 Supplier Mikromol Supplier No. MM1005.06
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(14bRS)-2-Methyl-1,3,4,14b-tetrahydropyrazino[2,1-a]pyrido-[2,3-c][2]benzazepin-10(2H)-one (10-Oxomirtazapine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10-Oxo Mirtazapine
CAS 191546-97-1
Analyte or Component Names 10-Oxo Mirtazapine
Shipping Condition Room Temperature
Molecular Formula C17 H17 N3 O
Certifications/Compliance ISO 9001
Synonym Mirtazapine Imp. F (EP), Mirtazapine USP Related Compound D, Mirtazapine USP RC D, Pyrazino[2,1-a]pyrido[2,3-c][2]benzazepin-10(2H)-one, 1,3,4,14b-tetrahydro-2-methyl-, 2-Methyl-1,2,3,4-tetrahydrobenzo[c]pyrazino[1,2-a]pyrido[3,2-f]azepin-10(14bH)-one, Mirtazapine RC D (USP), Mirtazapine Related Compound D, Mirtazapine Impurity F
InChI Formula InChI=1S/C17H17N3O/c1-19-9-10-20-15(11-19)12-5-2-3-6-13(12)16(21)14-7-4-8-18-17(14)20/h2-8,15H,9-11H2,1H3
SMILES CN1CCN2C(C1)c3ccccc3C(=O)c4cccnc24
IUPAC Name (14bRS)-2-Methyl-1,3,4,14b-tetrahydropyrazino[2,1-a]pyrido-[2,3-c][2]benzazepin-10(2H)-one
Molecular Weight (g/mol) 279.34
Quantity 100 mg
Formula Weight 279.1372 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.