missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 142D6

Product Code. 30243660
Click to view available options
:
5 mg
This item is not returnable. View return policy

Product Code. 30243660

Brand: Tocris Bioscience™ 8018/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent Lys-covalent pan-IAP inhibitor

142D6 is a potent pan-IAP inhibitor (IC50 values are 21, 21 and 43 nM for cIAP1, cIAP2 and XIAP respectively). 142D6 covalently targets Lys residues in the BIR3 domain of the inhibitor of the apoptosis protein (IAP) family. In vitro, 142D6 reduces viability of cultured breast cancer MDA-MB-231 cells (EC50 values are 44 and 61 nM in cell viability and apoptosis assays, respectively). 142D6 is chemically stable in buffer and plasma and is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-1-((S)-1-((S)-2-Cyclohexyl-2-((S)-2-(methylamino)propanamido)acetyl)pyrrolidine-2-carboxamido)-2,3-dihydro-1H-inden-4-yl sulfurofluoridate
CAS 2410953-19-2
Quantity 5 mg
Target IAP Inhibitors
Molecular Formula C26H37FN4O6S
Purity 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.