missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 1400W dihydrochloride
GREENER_CHOICE

Product Code. 30001249
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001249

Brand: Tocris Bioscience™ 1415/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, highly selective iNOS inhibitor

Slow, tight binding, potent and highly selective inhibitor of inducible nitric oxide synthase (Kd = 7 nM). Selective over nNOS and eNOS (Ki values are 2 and 50 μM respectively). Cell-permeable and active in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214358-33-5
Molecular Formula C10H17Cl2N3

Specifications

Quantity 10 mg
Formula Weight 250.17
Purity >99%
Molecular Weight (g/mol) 250.167
Chemical Name or Material 1400W dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.