missing translation for 'onlineSavingsMsg'
Learn More

14-Deoxy-11,12-didehydroandrographolide, MedChemExpress

Product Code. 16441648
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16441648

Brand: Medchem Express HYN1490

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide. 14-Deoxy-11,12-didehydroandrographolide inhibits NF-κB activation.

14-Deoxy-11,12-didehydroandrographolide is an analogue of Andrographolide. 14-Deoxy-11,12-didehydroandrographolide inhibits NF-κB activation.

Specifications

Chemical Name or Material 14-Deoxy-11,12-didehydroandrographolide
CAS 42895-58-9
Color White
Purity Grade Notes Research
Molecular Formula C20H28O4
Quantity 5 mg
Synonym 14-dehydro Andrographolide AP10
Solubility Information DMSO : 110 mg/mL (330.90 mM; Need ultrasonic) ∣DMF : 100 mg/mL (300.82 mM; Need ultrasonic)
SMILES O=C1OCC=C1/C=C/[C@@H]2C(CC[C@]3([H])[C@](C)(CO)[C@H](O)CC[C@@]23C)=C
Molecular Weight (g/mol) 332.43
Formula Weight 332.43
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.