Learn More
Invitrogen™ 14-3-3 theta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527641
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, H1299, HeLaS3, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (80%), Rabbit (99%), Chicken (99%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse and rat orthologs. This gene is upregulated in patients with amyotrophic lateral sclerosis. It contains in its 5' UTR a 6 bp tandem repeat sequence which is polymorphic, however, there is no correlation between the repeat number and the disease.
Specifications
| 14-3-3 theta | |
| Polyclonal | |
| Unconjugated | |
| YWHAQ | |
| 14-3-3; 14-3-3 protein tau; 14-3-3 protein T-cell; 14-3-3 protein theta; 14-3-3 tau; 14-3-3 theta; 14-3-3t; 1C5; 2700028P07Rik; AA409740; AU021156; HS1; OTTHUMP00000165887; Protein HS1; protein tau; protein, theta; R74690; tyrosine 3/tryptophan 5 -monooxygenase activation protein, theta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide; Unknown (protein for MGC:127486); YWHAQ | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10971, 22630 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.84 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P27348, P68254 | |
| YWHAQ | |
| Recombinant protein encompassing a sequence within the center region of human 14-3-3 theta. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.