missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 tau/theta Antibody (1A1), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
14-3-3 tau/theta Monoclonal antibody specifically detects 14-3-3 tau/theta in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Formulation | 50mM Sodium Borate |
| Gene Alias | 14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide |
| Host Species | Mouse |
| Immunogen | Recombinant human 14-3-3 tau/theta (1-245aa) purified from E. coli (NP_006817). |
| Quantity | 0.1 mL |
| Target Species | Human |
| Isotype | IgG1 κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?