missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 tau/theta Antibody (1A1), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-21301AF532
This item is not returnable.
View return policy
Description
14-3-3 tau/theta Monoclonal specifically detects 14-3-3 tau/theta in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| 14-3-3 tau/theta | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| YWHAQ | |
| Recombinant human 14-3-3 tau/theta (1-245aa) purified from E. coli (NP_006817). | |
| 0.1 mL | |
| 10971.0 | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1A1 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction