missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 sigma/Stratifin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98309-50ul
This item is not returnable.
View return policy
Description
14-3-3 sigma/Stratifin Polyclonal antibody specifically detects 14-3-3 sigma/Stratifin in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| 14-3-3 sigma/Stratifin | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:2000-1:20000 | |
| 14-3-3 protein sigma, 14-3-3 sigma, Epithelial cell marker protein 1, HME1, stratifin, YWHAS | |
| Produced in rabbits immunized with purified, recombinant Mouse 14-3-3 sigma/Stratifin (Accession#: NP_061224.2; Met1-Ser248). | |
| 50 μg | |
| Cell Cycle and Replication | |
| 2810 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction