Learn More
Invitrogen™ 14-3-3 sigma Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585350
Description
Keep as concentrated solution. Predicted reactivity: Mouse (98%), Pig (97%), Chicken (81%), Sheep (97%), Rhesus Monkey (99%), Bovine (97%). Positive Control: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a cell cycle checkpoint protein. The encoded protein binds to translation and initiation factors and functions as a regulator of mitotic translation. In response to DNA damage this protein plays a role in preventing DNA errors during mitosis.
Specifications
| 14-3-3 sigma | |
| Polyclonal | |
| Unconjugated | |
| SFN | |
| 14-3-3 protein sigma; 14-3-3 sigma; 14-3-3 sigma protein; 14-3-3sigma; epithelial cell marker protein 1; Er; HGNC:10773; HME1; Mkrn3; Mme1; SFN; Stratifin; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, sigma polypeptide; Ywhas | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2810 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P31947 | |
| SFN | |
| Recombinant protein encompassing a sequence within the center region of human 14-3-3 sigma. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.