missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 Sigma Mouse anti-Human, CoraLite 650, Clone: 3F10D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL650-66251-100UL
This item is not returnable.
View return policy
Description
14-3-3 Sigma Monoclonal antibody apecifically detetects 14-3-3 Sigma antigen in Human Samples. It is validated for Flow Cytometry.Specifications
| 14-3-3 Sigma | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| SFN | |
| 14-3-3 Sigma fusion protein Ag12644 | |
| 100 μL | |
| 2810 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Flow Cytometry | |
| 3F10D9 | |
| CoraLite 650 | |
| BC002995 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction