Learn More
14-3-3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14503-1-AP-20UL
 View more versions of this product
Product Code. 16893164
  • £135.00 / 20µL
Description
This antibody was raised against full-length 14-3-3 theta.
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse and rat orthologs. This gene is upregulated in patients with amyotrophic lateral sclerosis. It contains in its 5′ UTR a 6 bp tandem repeat sequence which is polymorphic, however, there is no correlation between the repeat number and the disease.
Specifications
Specifications
14-3-3
Polyclonal
Unconjugated
YWHAQ
14 3 3, 14 3 3 protein T cell, 14 3 3 protein tau, 14 3 3 protein theta, 1C5, HS1, Protein HS1, YWHAQ
Rabbit
Antigen Affinity Chromatography
RUO
10971, 22630, 25577
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Flow Cytometry
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P27348, P68254, P68255
Ywhaq
14-3-3 Fusion Protein Ag5959
20 μL
Primary
Mouse, Rat, Human