missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14503-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody was raised against full-length 14-3-3 theta.
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse and rat orthologs. This gene is upregulated in patients with amyotrophic lateral sclerosis. It contains in its 5′ UTR a 6 bp tandem repeat sequence which is polymorphic, however, there is no correlation between the repeat number and the disease.Specifications
| 14-3-3 | |
| Polyclonal | |
| Unconjugated | |
| YWHAQ | |
| 14 3 3, 14 3 3 protein T cell, 14 3 3 protein tau, 14 3 3 protein theta, 1C5, HS1, Protein HS1, YWHAQ | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10971, 22630, 25577 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27348, P68254, P68255 | |
| Ywhaq | |
| 14-3-3 Fusion Protein Ag5959 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction