missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 14-3-3 Pan Recombinant Rabbit Monoclonal Antibody (2P9P6)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£147.00 - £363.00

Specifications

Antigen 14-3-3 Pan
Clone 2P9P6
Concentration 1.73 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283689
View Documents
Invitrogen™
MA558079
20 μL
£147.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282452
GREENER_CHOICE
View Documents
Invitrogen™
MA558080
100 μL
£363.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: MEKTELIQKA KLAEQAERYD DMATCMKAVT EQGAELSNEE RNLLSVAYKN VVGGRRSAWR VISSIEQKTD TSDKKLQLIK DYREKVESEL RSICTTVLEL.

At least seven isoforms comprise the highly conserved 14-3-3 family of homo- and heterodimeric proteins that are abundantly expressed in all eukaryotic cells. Although more than seven isoforms of 14-3-3 have been described, some redundancies have appeared upon sequencing. The 14-3-3s are thought to be key regulators of signal transduction events mediated through their binding to serine-phosphorylated proteins. By interacting with Cdc25C, 14-3-3 regulates entry into the cell cycle and, through interaction with Bad, prevents apoptosis. Other proteins that have been shown to bind to 14-3-3s include members of the protein kinase C family, Cbl, IRS-1, polyoma middle-T antigen, nitrate reductase, S-raf and the IGF-1 receptor. Detection of 14-3-3 proteins in cerebrospinal fluid has been shown to be quite useful in the differential diagnosis of Creutzfeldt-Jakob disease and other prion diseases.
Specifications

Specifications

14-3-3 Pan
1.73 mg/mL
ELISA, Western Blot
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
O70456, P27348, P31946, P31947, P35213, P61981, P61982, P61983, P62258, P62259, P62260, P63101, P63102, P63104, P68254, P68255, Q9CQV8
10971, 22627, 22628, 22630, 22631, 25577, 25578, 2810, 29753, 313017, 54401, 55948, 56010, 56011, 7529, 7531, 7532, 7534
A synthetic peptide corresponding to a sequence within amino acids human 1-100 of human pan 14-3-3 (NP_006817.1).
Antibody
2P9P6
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
YWHAB
1110013I11Rik; 1300003C17Rik; 14-3-3; 14-3-3 alpha; 14-3-3 beta; 14-3-3 delta; 14-3-3 epsilon; 14-3-3 gamma; 14-3-3 protein beta; 14-3-3 protein beta/alpha; 14-3-3 protein beta/alpha, N-terminally processed; 14-3-3 protein beta/alpha-A; 14-3-3 protein beta/alpha-B; 14-3-3 protein beta-subtype; 14-3-3 protein epsilon; 14-3-3 protein gamma; 14-3-3 protein gamma subtype; 14-3-3 protein gamma, N-terminally processed; 14-3-3 protein gamma-A; 14-3-3 protein gamma-B; 14-3-3 protein gamma-subtype; 14-3-3 protein sigma; 14-3-3 protein tau; 14-3-3 protein T-cell; 14-3-3 protein theta; 14-3-3 protein zeta; 14-3-3 protein zeta/delta; 14-3-3 protein/cytosolic phospholipase A2; 14-3-3 sigma; 14-3-3 sigma protein; 14-3-3 tau; 14-3-3 theta; 14-3-3 zeta; 14-3-3beta; 14-3-3d; 14-3-3E; 14-3-3G; 14-3-3gamma; 14-3-3-like protein; 14-3-3t; 14-3-3z; 14-3-3zeta; 14-3-3-zeta; 1C5; 2700028P07Rik; 3-monooxgenase/tryptophan 5-monooxgenase activation protein, gamma polypeptide; 3-monooxgenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; 3-monooxygenase/tryptophan 5 monooxygenase activation protein beta polypeptide; 3-monooxygenase/tryptophan 5-monooxgenase activation protein gamma polypeptide; 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; AA409740; AI596267; AL022924; AU019196; AU020854; AU021156; brain protein 14-3-3, beta isoform; D7Bwg1348e; epididymis luminal protein 2; epididymis luminal protein 4; epididymis secretory protein Li 1; epididymis secretory protein Li 3; epididymis secretory protein Li 93; epithelial cell marker protein 1; Er; factor activating exoenzyme S; FAS; GW128; HEL2; HEL4; HEL-S-1; HEL-S-3; HEL-S-93; HME1; HS1; KCIP-1; MDCR; MDS; mitochondrial import stimulation factor (MSF) L subunit; mitochondrial import stimulation factor L subunit; Mitochondrial import stimulation factor S1 subunit; Mkrn3; Mme1; MSF L; Msfs1; phospholipase A2; PPP1R170; prepronerve growth factor RNH-1; Protein 1054; Protein HS1; Protein kinase C inhibitor protein 1; protein kinase C inhibitor protein-1; protein phosphatase 1, regulatory subunit 170; protein tau; protein, theta; R74690; SEZ-2; SFN; Stratifin; tryosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein gamma; tryosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein gamma polypeptide; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta polypeptide; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma; tyrosine 3/tryptophan 5 -monooxygenase activation protein beta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein gamma polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, beta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, gamma polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, theta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, zeta polypeptide; tyrosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein, beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein, beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein, gamma polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta L homeolog; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta S homeolog; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma L homeolog; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma S homeolog
Sfn, YWHAB, YWHAE, YWHAG, YWHAQ, YWHAZ
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.