missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 gamma Antibody (505810) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30648411 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30648411 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30648411 Supplier Novus Biologicals Supplier No. FAB5700JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

14-3-3 gamma Monoclonal antibody specifically detects 14-3-3 gamma in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 gamma
Applications Western Blot
Classification Monoclonal
Clone 505810
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias 14-3-3 gamma, 14-3-3 protein gamma, 14-3-3GAMMA, KCIP-1, Protein kinase C inhibitor protein 1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gammapolypeptide
Host Species Mouse
Immunogen E. coli-derived recombinant human 14-3-3 gamma , Met1-Asn247, Accession # P61981
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7532
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.