missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 eta Antibody, FITC, Novus Biologicals™

Product Code. 18342464 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342464 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342464 Supplier Novus Biologicals Supplier No. NBP299642F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

14-3-3 eta Polyclonal specifically detects 14-3-3 eta in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 eta
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias 14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant 14-3-3 eta (Q04917; Gly2-Asn246).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7533
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.