missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 eta Antibody (3G12), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18404926 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404926 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404926 Supplier Novus Biologicals Supplier No. NBP192691JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

14-3-3 eta Monoclonal antibody specifically detects 14-3-3 eta in Human, Mouse, Rat, Bovine, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 eta
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Gene Knock-Out
Classification Monoclonal
Clone 3G12
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias 14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta
Host Species Mouse
Immunogen Full length recombinant human 14-3-3 eta expressed in and purified from E.coli [UniProt# Q04917].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7533
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.