missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
14-3-3 eta Monoclonal antibody specifically detects 14-3-3 eta in Human, Mouse, Rat, Bovine, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, KnockDown
Specifications
Specifications
| Antigen | 14-3-3 eta |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown |
| Classification | Monoclonal |
| Clone | 3G12 |
| Conjugate | DyLight 594 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Knockout Validated |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta |
| Host Species | Mouse |
| Immunogen | Full length recombinant human 14-3-3 eta expressed in and purified from E.coli [UniProt# Q04917]. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?