missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 epsilon Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11648-2-AP-150UL
This item is not returnable.
View return policy
Description
This antibody was raised against full-length 14-3-3 epsilon.
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the mouse ortholog. It interacts with CDC25 phosphatases, RAF1 and IRS1 proteins, suggesting its role in diverse biochemical activities related to signal transduction, such as cell division and regulation of insulin sensitivity. It has also been implicated in the pathogenesis of small cell lung cancer. Two transcript variants, one protein-coding and the other non-protein-coding, have been found for this gene.Specifications
| 14-3-3 epsilon | |
| Polyclonal | |
| Unconjugated | |
| YWHAE | |
| 14 3 3 protein epsilon, 14 3 3E, 14-3-3, 14-3-3 epsilon, KCIP 1, MDCR, MDS, YWHAE | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22627, 29753, 7531 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62258, P62259, P62260 | |
| YWHAE | |
| 14-3-3 epsilon Fusion Protein Ag2247 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction