missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 epsilon Rabbit anti-Human, Mouse, Rat, Clone: 4X6E1, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | 14-3-3 epsilon |
|---|---|
| Clone | 4X6E1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18359421
|
Novus Biologicals
NBP3-16515-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18370474
|
Novus Biologicals
NBP3-16515-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
14-3-3 epsilon Monoclonal antibody specifically detects 14-3-3 epsilon in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| 14-3-3 epsilon | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Lipid and Metabolism | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 7531 | |
| IgG | |
| Affinity purified |
| 4X6E1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 14-3-3 epsilon, 14-3-3 protein epsilon, FLJ45465, FLJ53559,14-3-3E, KCIP-1, MDCR, MDS, mitochondrial import stimulation factor L subunit, protein kinase C inhibitor protein-1, tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilonpolypeptide | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human 14-3-3 epsilon (P62258). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title