missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 beta Antibody (05), Janelia Fluor™ 646 , Novus Biologicals™

Product Code. 18418969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418969 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418969 Supplier Novus Biologicals Supplier No. NBP306157JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

14-3-3 beta Monoclonal antibody specifically detects 14-3-3 beta in Cynomolgus Monkey samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 beta
Applications Western Blot, ELISA
Classification Monoclonal
Clone 05
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias 14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Cynomolgus 14-3-3 beta (Q4R572-2; Met 2-Asn 244).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7529
Target Species Cynomolgus Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.