missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 beta/alpha Rabbit anti-Human, Mouse, Rat, Clone: S08-4A7, Novus Biologicals™

Product Code. 18379703
Change view
Click to view available options
Quantity:
100 μL
25 ul
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379703 25 ul 25µL
18371347 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18379703 Supplier Bio-Techne Supplier No. NBP31490625UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

14-3-3 beta/alpha Monoclonal antibody specifically detects 14-3-3 beta/alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 beta/alpha
Applications Immunohistochemistry
Classification Monoclonal
Clone S08-4A7
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias 14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha, brain protein 14-3-3, beta isoform, GW128, HS1, KCIP-1, Protein 1054, Protein kinase C inhibitor protein 1, protein kinase C inhibitor protein-1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, alphapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, betapolypeptide, YWHAA
Host Species Rabbit
Immunogen A synthetic peptide of human 14-3-3 beta/alpha (Uniprot#: P31946)
Purification Method Affinity purified
Quantity 25 ul
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7529
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.