missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | 14-3-3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-4ug/ml, Simple Western 1:10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18175312
|
Novus Biologicals
NBP2-27370SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18104625
|
Novus Biologicals
NBP2-27370 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
14-3-3 Polyclonal specifically detects 14-3-3 in Human, Primate samples. It is validated for Western Blot, Simple Western.Specifications
| 14-3-3 | |
| Western Blot 2-4ug/ml, Simple Western 1:10 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Primate | |
| P27348 | |
| 10971 | |
| A portion of amino acids 40-90 from human 14-3-3 was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide | |
| YWHAQ | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, crab-eating macaque, and rhesus monkey. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title