Learn More
Epredia™ Lab Vision™ 14.3.3σ Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Take advantage of a quality product that delivers accurate results with the Epredia™ 14.3.3σ Ab-1, Mouse Monoclonal Antibody.
£108.12 - £601.80
Specifications
| Antigen | 14.3.3sigma Ab-1 |
|---|---|
| Clone | 1433S01 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|
12619236
|
Epredia™
MS-1185-P0 |
100 μL | Concentrated | 200μg/mL | |||||
|
12639236
|
Epredia™
MS-1185-P1ABX |
100 μL | Concentrated | 1mg/mL | |||||
|
12649236
|
Epredia™
MS-1185-PABX |
200 μL | Concentrated | 1mg/mL | |||||
|
12629236
|
Epredia™
MS-1185-P1 |
500 μL | Concentrated | 200μg/mL | |||||
|
12609236
|
Epredia™
MS-1185-P |
1 mL | Concentrated | 200μg/mL | |||||
|
12689236
|
Epredia™
MS-1185-R7 |
7 mL | Prediluted | Prediluted | |||||
Description
14.3.3 proteins are a family of small, widely expressed, highly conserved cytosolic proteins. 14.3.3 proteins bind to and influence the activities of a diverse group of molecules involved in signal transduction, cell cycle regulation and apoptosis, including Raf, PKC, Bad, Cbl, and c-Bcr. Interactions between 14.3.3 and target proteins are strongly influenced by the phosphorylation state of 14.3.3 and the target protein. 14.3.3 σ shares cyclin-Cdk2 binding motifs with different cell cycle regulators, including p107, p130, p21Cip1, p27Kip1, and p57Kip2 and is associated with cyclin-Cdk complexes in vitro and in vivo.Host Species: Mouse
Clone: 1433S01
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant human 14.3.3σ protein
Molecular Weight: 26-30kDa
Positive Control: HT29 cells, Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
Note:
USA: RUO; Int'l: RUO
Specifications
| 14.3.3sigma Ab-1 | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| IgG2a κ |
| 1433S01 | |
| Monoclonal | |
| Mouse | |
| Signal Transduction | |
| Recombinant human 1433 sigma protein | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.