missing translation for 'onlineSavingsMsg'
Learn More

13Beta-Ethyl-11-methyliden-18.19-dinor-3-ethynyl-pregna-2,4-diene-17-one, TRC

Product Code. 30384617
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384617 2.5 mg 2.5mg
30383159 1 mg 1mg
30370379 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384617 Supplier Toronto Research Chemicals Supplier No. TRCE9336702.5MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 13beta-Ethyl-11-methyliden-18.19-dinor-3-ethynyl-pregna-2,4-diene-17-one
Analyte or Component Names 13beta-Ethyl-11-methyliden-18.19-dinor-3-ethynyl-pregna-2,4-diene-17-one
Shipping Condition Room Temperature
Molecular Formula C22H26O
Synonym (8S,9S,10R,13S,14S)-13-Ethyl-3-ethynyl-11-methylene-7,8,9,10,11,12,13,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17(6H)-one
InChI Formula InChI=1S/C22H26O/c1-4-15-6-8-17-16(12-15)7-9-18-19-10-11-20(23)22(19,5-2)13-14(3)21(17)18/h1,6,12,17-19,21H,3,5,7-11,13H2,2H3/t17-,18-,19-,21+,22-/m0/s1
Solution Type Neat
SMILES C=C1C[C@@]([C@](CC2)([H])[C@@](CC3)([H])[C@@]1([H])[C@]4([H])C3=CC(C#C)=CC4)(C2=O)CC
IUPAC Name (8S,9S,10R,13S,14S)-13-ethyl-3-ethynyl-11-methylene-1,6,7,8,9,10,11,12,13,14,15,16-dodecahydro-17H-cyclopenta[a]phenanthren-17-one
Industry Type Forensics
Molecular Weight (g/mol) 306.44
Quantity 2.5 mg
Formula Weight 306.2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.