missing translation for 'onlineSavingsMsg'
Learn More

13Beta-Ethyl-11-methyliden-18.19-dinor-3,17Beta-dihydroxy-3-ethynyl-pregna-4-ene, TRC

Product Code. 30386117
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386117 25 mg 25mg
30371686 5 mg 5mg
30383513 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386117 Supplier Toronto Research Chemicals Supplier No. TRCE93367525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 13beta-Ethyl-11-methyliden-18.19-dinor-3,17beta-dihydroxy-3-ethynyl-pregna-4-ene
Analyte or Component Names 13beta-Ethyl-11-methyliden-18.19-dinor-3,17beta-dihydroxy-3-ethynyl-pregna-4-ene
Shipping Condition Room Temperature
Molecular Formula C22H30O2
Synonym (8S,9S,10R,13S,14S,17S)-13-Ethyl-3-ethynyl-11-methylene-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol
InChI Formula InChI=1S/C22H30O2/c1-4-21(24)11-10-16-15(13-21)6-7-17-18-8-9-19(23)22(18,5-2)12-14(3)20(16)17/h1,13,16-20,23-24H,3,5-12H2,2H3/t16-,17-,18-,19-,20+,21?,22-/m0/s1
Solution Type Neat
SMILES C=C1C[C@@]([C@]2([H])[C@@](CC3)([H])[C@@]1([H])[C@]4([H])C3=CC(O)(C#C)CC4)([C@@H](O)CC2)CC
IUPAC Name (8S,9S,10R,13S,14S,17S)-13-ethyl-3-ethynyl-11-methylene-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,17-diol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 326.47
Quantity 25 mg
Formula Weight 326.22
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.