missing translation for 'onlineSavingsMsg'
Learn More

(13Alpha,14Alpha)-14,19-Dihydro-12,13-[methylenebis(oxy)]-20-norcrotalanan-11,15-dione, TRC

Product Code. 30386324
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386324 2.5 g -
30377555 1 g -
30388504 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386324 Supplier Toronto Research Chemicals Supplier No. TRCD4496432.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121719-98-0
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.37
Synonym 7H-[1,3]Dioxolo[8,9][1,6]dioxacycloundecino[2,3,4-gh]pyrrolizine, 20-norcrotalanan-11,15-dione Derivative
SMILES C[C@H]1C(=O)O[C@@H]2CCN3CC=C(COC(=O)[C@]4(C)OCO[C@]14C)[C@H]23

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C17H23NO6/c1-10-14(19)24-12-5-7-18-6-4-11(13(12)18)8-21-15(20)17(3)16(10,2)22-9-23-17/h4,10,12-13H,5-9H2,1-3H3/t10-,12+,13+,16+,17-/m0/s1
Formula Weight 337.1525
Chemical Name or Material (13alpha,14alpha)-14,19-Dihydro-12,13-[methylenebis(oxy)]-20-norcrotalanan-11,15-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.