missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Bouillon De Base Fraser
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
Description
Selectively enrich and isolate Listeria spp. from food and environmental samples with Fraser broth Base.
Specifications
Specifications
| Format | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 8.7 L medium |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction