missing translation for 'onlineSavingsMsg'
Learn More

13-O-Acetyl-9-dihydrobaccatin III, Mikromol™

Product Code. 30050279
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30050279

Brand: Mikromol MM1489.120025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

13-O-Acetyl-9-dihydrobaccatin III
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 13-acetyl-9-dihydrobaccatin III
CAS 142203-65-4
Analyte or Component Names 13-acetyl-9-dihydrobaccatin III
Shipping Condition Room Temperature
Molecular Formula C33 H42 O12
Certifications/Compliance ISO 17025
Synonym 13-O-Acetyl-9-dihydrobaccatin III, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,5,6,9,11,12,12b-heptol, 2a,3,4,4a,5,6,9,10,12,12a-decahydro-4a,8,13,13-tetramethyl-, 6,9,12b-triacetate 12-benzoate, (2aR,4S,4aS,5R,6R,9S,11S,12S,12aR,12bS)-, 13-Acetyl-9-dihydrobaccatin III, 7,9-dideacetylbaccatin VI, 9-Dihydro-13-acetylbaccatin III, Di-O-deacetylbaccatin VI, [2aR-(2alpha,4beta,4abeta,5alpha,6beta,9alpha,11alpha,12alpha,12aalpha,12balpha)]-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,5,6,9,11,12,12b-heptol, 2a,3,4,4a,5,6,9,10,12,12a-decahydro-4a,8,13,13-tetramethyl-, 6,9,12b-triacetate 12-benzoate, [2aR-(2aalpha,4beta,4abeta,5alpha,6beta,9alpha,11alpha,12alpha,12aalpha,12balpha)]-
InChI Formula InChI=1S/C33H42O12/c1-16-21(42-17(2)34)14-33(40)28(44-29(39)20-11-9-8-10-12-20)26-31(7,22(37)13-23-32(26,15-41-23)45-19(4)36)27(38)25(43-18(3)35)24(16)30(33,5)6/h8-12,21-23,25-28,37-38,40H,13-15H2,1-7H3/t21-,22-,23+,25+,26-,27-,28-,31+,32-,33+/m0/s1
SMILES CC(=O)O[C@H]1C[C@@]2(O)[C@@H](OC(=O)c3ccccc3)[C@H]4[C@@](C)([C@@H](O)C[C@H]5OC[C@@]45OC(=O)C)[C@@H](O)[C@H](OC(=O)C)C(=C1C)C2(C)C
IUPAC Name (2aR,4S,4aS,5R,6R,9S,11S,12S,12aR,12bS)-2a,3,4,4a,5,6,9,10,12,12a-decahydro-4a,8,13,13-tetramethyl-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,5,6,9,11,12,12b-heptol 6,9,12b-triacetate 12-benzoate
Molecular Weight (g/mol) 630.68
Quantity 25 mg
Formula Weight 630.2676 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.