missing translation for 'onlineSavingsMsg'
Learn More

13-Ethyl-17-hydroxy-18,19-dinor-17Alpha-pregn-5(10)-en-20-yn-3-one(Levo Norgestrel Impurity), TRC

Product Code. 30377151
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377151 25 mg
30388406 5 mg
30370667 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377151 Supplier Toronto Research Chemicals Supplier No. TRCE91893525MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19914-67-1
Molecular Formula C21 H28 O2
Molecular Weight (g/mol) 312.45
Synonym (17α)-13-ethyl-17-hydroxy-18,19-Dinorpregn-5(10)-en-20-yn-3-one, 18,19-Dinor-17α-pregn-5(10)-en-20-yn-3-one, 13-ethyl-17-hydroxy- (7CI,8CI), 13-Ethyl-17α-ethynyl-17-hydroxygon-5(10)-en-3-one, Levonorgestrel Imp. B (EP)
IUPAC Name (8R,9S,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-1,2,4,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC[C@]12CC[C@H]3[C@@H](CCC4=C3CCC(=O)C4)[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,17-19,23H,3,5-13H2,1H3/t17-,18-,19+,20+,21+/m1/s1
Formula Weight 312.2089
Chemical Name or Material 13-Ethyl-17-hydroxy-18,19-dinor-17a-pregn-5(10)-en-20-yn-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.