missing translation for 'onlineSavingsMsg'
Learn More
Learn More
13-Acetyl-9-dihydrobaccatin-III, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | 13-acetyl-9-dihydrobaccatin III |
| CAS | 142203-65-4 |
| Molecular Formula | C33 H42 O12 |
| Quantity | 5 mg |
| Synonym | 13-O-Acetyl-9-dihydrobaccatin III, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,5,6,9,11,12,12b-heptol, 2a,3,4,4a,5,6,9,10,12,12a-decahydro-4a,8,13,13-tetramethyl-, 6,9,12b-triacetate 12-benzoate, (2aR,4S,4aS,5R,6R,9S,11S,12S,12aR,12bS)-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,5,6,9,11,12,12b-heptol, 2a,3,4,4a,5,6,9,10,12,12a-decahydro-4a,8,13,13-tetramethyl-, 6,9,12b-triacetate 12-benzoate, [2aR-(2aα,4β,4aβ,5α,6β,9α,11α,12α,12aα,12bα)]-, 13-Acetyl-9-dihydrobaccatin III, 7,9-dideacetylbaccatin VI, 9-Dihydro-13-acetylbaccatin III, Di-O-deacetylbaccatin VI |
| InChI Formula | InChI=1S/C33H42O12/c1-16-21(42-17(2)34)14-33(40)28(44-29(39)20-11-9-8-10-12-20)26-31(7,22(37)13-23-32(26,15-41-23)45-19(4)36)27(38)25(43-18(3)35)24(16)30(33,5)6/h8-12,21-23,25-28,37-38,40H,13-15H2,1-7H3/t21-,22-,23+,25+,26-,27-,28-,31+,32-,33+/m0/s1 |
| SMILES | CC(=O)O[C@H]1C[C@@]2(O)[C@@H](OC(=O)c3ccccc3)[C@H]4[C@@](C)([C@@H](O)C[C@H]5OC[C@@]45OC(=O)C)[C@@H](O)[C@H](OC(=O)C)C(=C1C)C2(C)C |
| IUPAC Name | (2aR,4S,4aS,5R,6R,9S,11S,12S,12aR,12bS)-2a,3,4,4a,5,6,9,10,12,12a-decahydro-4a,8,13,13-tetramethyl-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,5,6,9,11,12,12b-heptol 6,9,12b-triacetate 12-benzoate |
| Molecular Weight (g/mol) | 630.68 |
| Formula Weight | 630.2676 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction