missing translation for 'onlineSavingsMsg'
Learn More

LEC Refrigeration Refrigerator PE907 pharmacy 286L 595mm(w) x 600mm(d) x 1550mm(h)

Brand:  LEC Refrigeration PE907

Refrigerator PE907 pharmacy 286L 595mm(w) x 600mm(d) x 1550mm(h)

Code : 04

Additional Details : Weight : 81.50000kg

Product Code. 12938684

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.