missing translation for 'onlineSavingsMsg'
Learn More

LEC Refrigeration Refrigerator PE507 pharmacy 153L 558mm(w) x 600mm(d) x 865mm(h)

Brand:  LEC Refrigeration PE507

Refrigerator PE507 pharmacy 153L 558mm(w) x 600mm(d) x 865mm(h)

Code : 04

Additional Details : Weight : 34.00000kg

Product Code. 12928684

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.